543004
1,2-Diaminoethane trityl, polymer-bound
100-200 mesh, extent of labeling: 1.2-1.7 mmol/g loading, 1 % cross-linked
Synonym(s):
1,2-Diaminoethane trityl polystyrene resin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
crosslinking
1 % cross-linked
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
extent of labeling
1.2-1.7 mmol/g loading
particle size
100-200 mesh
functional group
amine
SMILES string
C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.NCCNC(c3ccccc3)(c4ccccc4)c5ccccc5
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of clinical endocrinology and metabolism, 91(2), 535-538 (2005-11-10)
The melanocortin 4 receptor (MC4R) is an essential regulator of energy intake and body weight. Recently, the V103I polymorphism of MC4R has been shown to be negatively associated with body mass index. This suggests that serum lipids and blood pressure
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service