Skip to Content
Merck
All Photos(1)

Key Documents

541788

Sigma-Aldrich

Diethyl 2-phenylethyl phosphonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)2PO(OC2H5)2
CAS Number:
Molecular Weight:
242.25
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.495 (lit.)

bp

125 °C/0.25 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CCc1ccccc1)OCC

InChI

1S/C12H19O3P/c1-3-14-16(13,15-4-2)11-10-12-8-6-5-7-9-12/h5-9H,3-4,10-11H2,1-2H3

InChI key

WEXGRJPJMNUCMG-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of 1,3-diketones, β-keto esters, and β-keto phosphonates via 1,2-addition of carbanionic nucleophiles to cyclic vinylogous acyl triflates, followed by ring-opening fragmentation, Claisen condensation and carbon-carbon bond cleavage
  • Controlling the surface state emission in ultrasmall CdSe nanocrystals
  • Reactions with organoboranes for synthesis of organophosphorous compounds

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service