Recommended Products
Assay
98%
mp
38-42 °C (lit.)
SMILES string
Fc1cccc(c1CBr)C(F)(F)F
InChI
1S/C8H5BrF4/c9-4-5-6(8(11,12)13)2-1-3-7(5)10/h1-3H,4H2
InChI key
RINUERVPFANASB-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
225.0 °F - closed cup
Flash Point(C)
107.2 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service