Skip to Content
Merck
All Photos(1)

Key Documents

538698

Sigma-Aldrich

3′-(Trifluoromethyl)propiophenone

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€63.30

€63.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€63.30

About This Item

Linear Formula:
CF3C6H4COCH2CH3
CAS Number:
Molecular Weight:
202.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€63.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.4620 (lit.)

bp

216-217 °C (lit.)

density

1.202 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CCC(=O)c1cccc(c1)C(F)(F)F

InChI

1S/C10H9F3O/c1-2-9(14)7-4-3-5-8(6-7)10(11,12)13/h3-6H,2H2,1H3

InChI key

UBTPNKXHYILGJU-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service