Skip to Content
Merck
All Photos(1)

Key Documents

537640

Sigma-Aldrich

Diethylene glycol butyl ether

≥99.2%

Synonym(s):

DB Solvent, 2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739225
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.6 (vs air)

vapor pressure

30 mmHg ( 130 °C)

product line

ReagentPlus®

Assay

≥99.2%

autoignition temp.

400 °F

expl. lim.

6.2 %

refractive index

n20/D 1.432 (lit.)

bp

231 °C (lit.)

mp

−68 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCOCCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E D Thompson et al.
Environmental health perspectives, 57, 105-112 (1984-08-01)
The mutagenic potential of diethylene glycol monobutyl ether (diEGBE) was examined with a Tier I battery of in vitro assays followed by a Tier II in vivo Drosophila sex-linked recessive lethal assay. The in vitro battery consisted of: the Salmonella
Reinhard B Raggam et al.
Mycoses, 58(7), 394-398 (2015-05-12)
Serum 1,3-beta-d-glucan (BDG) testing is an established diagnostic marker for invasive fungal infections (IFI) among patients with haematological malignancies. In contrast limited data exist regarding the application of urine BDG testing. Same-day midstream urine and serum screening samples were collected
M M Udden
Toxicology letters, 156(1), 95-101 (2005-02-12)
The toxicity of diethylene glycol butyl ether (DGBE), and its principal metabolite, butoxyethoxyacetic acid (BEAA), were assessed in vitro for rat and human red blood cells. Rat erythrocytes showed evidence of mild hemolysis when exposed to BEAA at concentrations of
Hypersensitivity to 2-(2-butoxyethoxy)ethanol.
K Berlin et al.
Contact dermatitis, 32(1), 54-54 (1995-01-01)
K A Johnson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(3), 467-481 (2005-02-01)
Standard toxicologic endpoints, supplemented by additional examinations, were studied for groups of 10 Fischer 344 rats/sex given drinking water formulated to supply 0, 50, 250, or 1000 mg diethylene glycol monobutyl ether (DGBE)/kg/day for 13 weeks. These dose levels were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service