Skip to Content
Merck
All Photos(1)

Documents

532916

Sigma-Aldrich

(S)-(+)-2-Amino-4-phenylbutyric acid ethyl ester hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)2CH(NH2)CO2C2H5 · HCl
CAS Number:
Molecular Weight:
243.73
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +26°, c = 1 in chloroform

reaction suitability

reaction type: solution phase peptide synthesis

mp

159-163 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)[C@@H](N)CCc1ccccc1

InChI

1S/C12H17NO2.ClH/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10;/h3-7,11H,2,8-9,13H2,1H3;1H/t11-;/m0./s1

InChI key

PTFKZMFFSIYCOV-MERQFXBCSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service