Skip to Content
Merck
All Photos(1)

Key Documents

531820

Sigma-Aldrich

N,N′-Di-Boc-thiourea

97%

Synonym(s):

N,N′−Di-(tert-butoxycarbonyl)thiourea, N,N′-Bis-tert-butoxycarbonylthiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3COCONH)2CS
CAS Number:
Molecular Weight:
276.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

131-135 °C (lit.)

functional group

amine
thiourea

SMILES string

CC(C)(C)OC(=O)NC(=S)NC(=O)OC(C)(C)C

InChI

1S/C11H20N2O4S/c1-10(2,3)16-8(14)12-7(18)13-9(15)17-11(4,5)6/h1-6H3,(H2,12,13,14,15,18)

InChI key

CSOJECDGWHHWRS-UHFFFAOYSA-N

General description

N,N′-Di-Boc-thiourea (N,N′-di-Boc-substituted thiourea), also known as di-Boc-thiourea is a thioacylating agent for preparing thiocarbonyl compounds.

Application

N,N′-Di-Boc-thiourea (di-Boc protected thiourea) may be used as a guanylating agent for the synthesis of guanidines, via guanylation of amines in the presence of Mukaiyama′s reagent as a promoter.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent developments in guanylating agents.
Katritzky AR and Rogovoy BV.
ARKIVOC (Gainesville, FL, United States), 4, 49-87 (2005)
N, N'-Di-Boc-substituted thiourea as a novel and mild thioacylating agent applicable for the synthesis of thiocarbonyl compounds.
Yin BL, et al.
Synthesis, 2010(06), 991-999 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service