Skip to Content
Merck
All Photos(1)

Key Documents

529923

Sigma-Aldrich

Butylmagnesium chloride solution

1.45 M in butyl diglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3MgCl
CAS Number:
Molecular Weight:
116.87
Beilstein:
3587228
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

1.45 M in butyl diglyme

density

0.945 g/mL at 25 °C

SMILES string

CCCC[Mg]Cl

InChI

1S/C4H9.ClH.Mg/c1-3-4-2;;/h1,3-4H2,2H3;1H;/q;;+1/p-1

InChI key

UUNGXYVLXYGZKH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Pons et al.
Journal of chromatography. B, Biomedical sciences and applications, 716(1-2), 139-145 (1998-11-21)
One analytical procedure for the determination of ionic alkyllead in human urine has been studied. The system consists of the extraction of Me3Pb+, Et3Pb+ and Pb2+ at pH 9.0 with diethyldithiocarbamate to an organic phase. Then, the ionic compounds are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service