Skip to Content
Merck
All Photos(1)

Key Documents

528919

Sigma-Aldrich

2-Bromo-4,6-dichloroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(Cl)2NH2
CAS Number:
Molecular Weight:
240.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

mp

81-83 °C (lit.)

SMILES string

Nc1c(Cl)cc(Cl)cc1Br

InChI

1S/C6H4BrCl2N/c7-4-1-3(8)2-5(9)6(4)10/h1-2H,10H2

InChI key

DTPADCOGQUOGHT-UHFFFAOYSA-N

General description

Bromo-4,6-dichloroaniline can be prepared from 2,4-dichloroacetanilide via deprotection, followed by bromination with NBS (N-bromosuccinimide).[1]

Application

2-Bromo-4,6-dichloroaniline reacts with Pd(OAc)2, PPh3, potassium furan-2-yltrifluoroborate and K2CO3 to yied N-allyl-2,4-dichloro-6-(furan-2-yl)aniline.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Lovell Read et al.
The Journal of organic chemistry, 78(3), 1311-1316 (2013-01-12)
A novel and efficient synthesis of phenanthridines and aza analogues is reported. The key step is a microwave-mediated intramolecular Diels-Alder cyclization of o-furyl(allylamino)arenes. In the presence of a catalytic amount of acid, the DA-adduct reacts further to give the dihydrophenanthridines
An efficient synthesis of [13C6]-3, 5-dichloroaniline.
Latli B, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 51(7), 283-285 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service