Skip to Content
Merck
All Photos(3)

Key Documents

527513

Sigma-Aldrich

7-Bromo-1-heptene

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)5CH=CH2
CAS Number:
Molecular Weight:
177.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

refractive index

n20/D 1.466 (lit.)

bp

27 °C/1 mmHg (lit.)

density

1.162 g/mL at 25 °C (lit.)

functional group

alkyl halide
allyl
bromo

SMILES string

BrCCCCCC=C

InChI

1S/C7H13Br/c1-2-3-4-5-6-7-8/h2H,1,3-7H2

InChI key

GNYDYUQVALBGGZ-UHFFFAOYSA-N

General description

7-Bromo-1-heptene is a linear halogenated alkene.

Application

7-Bromo-1-heptene may be used to synthesize:
  • boc-L-2-amino-8-nonenoic acid (boc = tert-butyloxycarbonyl)[1]
  • 9-hept-6-enyloxy-5-hydroxymethyl-2-isopropyl-1-methyl-1,4,5,6-tetrahydro-2H-benzo[e][1,4] diazocin-3-one[2]
  • 8-hydroxy-15,15,15-trifluoro-1-pentadece[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.9 °F - closed cup

Flash Point(C)

58.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan P Kozikowski et al.
ChemMedChem, 4(7), 1095-1105 (2009-04-28)
A series of benzolactam compounds were synthesized, some of which caused a concentration-dependent increase in sAPPalpha and decrease in Abeta production in the concentration range of 0.1-10 microM. Moreover, some compounds showed neuroprotective effects in the 10-20 microM range in
G H Stoll et al.
Journal of lipid research, 32(5), 843-857 (1991-05-01)
An analogue of the long-chain fatty acid salt, sodium stearate, was synthesized in which the hydrogen atoms at carbons 2, 3, and 18 were replaced by fluorine. The key step in the synthesis was the addition of 3-iodo-2,2,3,3-tetrafluoropropanoic acid amide
Norikazu Nishino et al.
Bioorganic & medicinal chemistry, 16(1), 437-445 (2007-09-29)
Inhibitors of histone deacetylases (HDACs) are a promising class of anticancer agents that effect gene regulation. To know the interaction of aliphatic cap groups with HDACs, cyclic tetrapeptide and bicyclic peptide disulfide hybrids were synthesized without aromatic ring in their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service