Skip to Content
Merck
All Photos(1)

Key Documents

526835

Sigma-Aldrich

PSS-Octamethyl substituted

Synonym(s):

1,3,5,7,9,11,13,15-Octamethylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Octamethylsilsesquioxane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€72.30
25 G
€248.00

€72.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€72.30
25 G
€248.00

About This Item

Empirical Formula (Hill Notation):
C8H24O12Si8
CAS Number:
Molecular Weight:
536.95
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€72.30


Please contact Customer Service for Availability

Request a Bulk Order

mp

>350 °C (lit.)

Quality Level

SMILES string

C[Si]12O[Si]3(C)O[Si]4(C)O[Si](C)(O1)O[Si]5(C)O[Si](C)(O2)O[Si](C)(O3)O[Si](C)(O4)O5

InChI

1S/C8H24O12Si8/c1-21-9-22(2)12-25(5)14-23(3,10-21)16-27(7)17-24(4,11-21)15-26(6,13-22)19-28(8,18-25)20-27/h1-8H3

InChI key

SOQGBGSEJYZNPS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Could you let me know the purity of this PSS-Octamethyl substituted material? Thanks

    1 answer
    1. The purity of this product is not determined. FTIR confirms the identity of the compound.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service