Skip to Content
Merck
All Photos(1)

Key Documents

523739

Sigma-Aldrich

Phoxime resin

100-200 mesh, extent of labeling: 0.85 mmol/g Cl loading, 1 % cross-linked with divinylbenzene

Synonym(s):

O-Chloroformyl-4-nitrobenzophenone oxime polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

Phoxime

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: carboxyl reactive

extent of labeling

0.85 mmol/g Cl loading

particle size

100-200 mesh

functional group

oxime

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service