Skip to Content
Merck
All Photos(2)

Key Documents

523674

Sigma-Aldrich

4-(Trimethylsilyl)phenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC6H4B(OH)2
CAS Number:
Molecular Weight:
194.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95.0%

mp

173-178 °C (lit.)

SMILES string

C[Si](C)(C)c1ccc(cc1)B(O)O

InChI

1S/C9H15BO2Si/c1-13(2,3)9-6-4-8(5-7-9)10(11)12/h4-7,11-12H,1-3H3

InChI key

NRPZMSUGPMYBCQ-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana Hamer et al.
Electrophoresis, 39(8), 1048-1053 (2018-02-01)
Glycosylated hemoglobin (HbA1c) detection is performed routinely in hospitals as it is the most widespread confirmatory diagnosis of diabetes mellitus. Here we present a novel CE method for measuring HbA1c by introducing silica nanoparticles (NPs) modified with a boronic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service