Skip to Content
Merck
All Photos(1)

Key Documents

522996

Sigma-Aldrich

2-Amino-4-methoxy-6-methyl-1,3,5-triazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4O
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

258-261 °C (lit.)

SMILES string

COc1nc(C)nc(N)n1

InChI

1S/C5H8N4O/c1-3-7-4(6)9-5(8-3)10-2/h1-2H3,(H2,6,7,8,9)

InChI key

NXFQWRWXEYTOTK-UHFFFAOYSA-N

General description

2-Amino-4-methoxy-6-methyl-1,3,5-triazine is formed during the photocatalytic decomposition of sulfonylurea (Sus) herbicide. It is also formed as a thermal decomposition product of chlorsulfuron (a sulfonylurea herbicide) and has been determined by GC using nitrogen-phosphorus detection (NPD).

Application

2-Amino-4-methoxy-6-methyl-1,3,5-triazine may be used in the synthesis of thifensulfuron.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Thifensulfuron Using Bis (trichloromethyl) carbonate.
Yao RS, et al.
Chinese Journal of Applied Chemistry / Ying Yong Hua Xue, 19(7), 687-689 (2002)
Photocatalytic degradation of sulfonylurea herbicides in aqueous TiO 2.
Vulliet E, et al.
Applied Catalysis. B, Environmental, 38(2), 127-137 (2002)
Analysis of sulfonylurea herbicides by gas-liquid chromatography. 1. Formation of thermostable derivatives of chlorsulfuron and metsulfuron-methyl.
Klaffenbach P and Holland PT.
Journal of Agricultural and Food Chemistry, 41(3), 388-395 (1993)
Analysis of sulfonylurea herbicides by gas-liquid chromatography. 2. Determination of chlorsulfuron and metsulfuron-methyl in soil and water samples.
Klaffenbach P and Holland PT.
Journal of Agricultural and Food Chemistry, 41(3), 396-401 (1993)
Rajae Chahboune et al.
Rapid communications in mass spectrometry : RCM, 29(15), 1370-1380 (2015-07-07)
Sulfonylureas are among the most important class of antidiabetic and herbicides. Solar light excitation and Advanced Oxidation Processes may result in the formation of a wide array of products owing to the relative complex structure. These products, that should be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service