Skip to Content
Merck
All Photos(1)

Key Documents

521590

Sigma-Aldrich

1-Chloro-5-iodopentane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I(CH2)5Cl
CAS Number:
Molecular Weight:
232.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

SMILES string

ClCCCCCI

InChI

1S/C5H10ClI/c6-4-2-1-3-5-7/h1-5H2

InChI key

GUUHKOCYWBEGGX-UHFFFAOYSA-N

General description

1-Chloro-5-iodopentane is a halogenated hydrocarbon. 1,10-dichlorododecane is formed as a major product from the reduction of 1-chloro-5-iodohexane with electrogenerated nickel(I) salen [salen = N,N′-bis(salicylidene)ethylenediamine].[1] Its IR spectra in liquid and solid states have been studied.[2] It affords dimer radical cation on reaction with OH radicals.[3] Synthesis of 1-chloro-5-iodopentane starting from 1,5-dichloropentane has been reported.[4]

Application

1-Chloro-5-iodopentane may be used in the synthesis of 2-(tert-butyldimet hylsiloxy)-11-chloro-5-undecyne[5] and cis-7-dodecen-1-ol acetate.[6]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of two macrolide aggregation pheromones from the flat grain beetle, Cryptolestes pusillus (Schonherr).
Millar JC, et al.
The Journal of Organic Chemistry, 48(23), 4404-4407 (1983)
Insect sex attractants. VI. 7-dodecen-1-ol acetates and congeners.
N Green et al.
Journal of medicinal chemistry, 10(4), 533-535 (1967-07-01)
Conformational analysis of 1-chloro-4-iodobutane and 1-chloro-5-iodopentane.
Crowder G and Ali S.
Journal of Molecular Structure, 34(1), 47-53 (1976)
S Lindstedt et al.
Clinical chemistry, 22(8), 1330-1338 (1976-08-01)
Gas chromatography/mass spectrometry was used to identify a series of acids in urine and serum from a child who died 26 h after birth in severe metabolic acidosis with high lactate excretion. cis-5-Decene-1, 10-dioic acid and cis-5-dodencene-1, 12-dioic acid were
Transformation of OH-adduct of 1-chloro-4-iodobutane into intra-molecular radical cation in neutral aqueous solution.
Mohan H, et al.
Chemical Physics Letters, 300(3), 493-498 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service