Skip to Content
Merck
All Photos(3)

Documents

520624

Sigma-Aldrich

Platinum(IV) oxide hydrate

≥99.9%

Synonym(s):

Adams′ catalyst, Platinum dioxide hydrate, Platinum oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PtO2 · xH2O
CAS Number:
Molecular Weight:
227.08 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.9%

composition

Pt, 77-81%

reaction suitability

core: platinum
reagent type: catalyst

mp

450 °C (lit.)

SMILES string

O=[Pt]=O

InChI

1S/2O.Pt

InChI key

YKIOKAURTKXMSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenation and hydrogenolysis of various functional groups; used as dehydrogenation catalyst; used for selective oxidation of primary alcohols.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishimoto, S.-I.; Ohtani, B. et al
Journal of the American Chemical Society, 105, 7180-7180 (1983)
Lee, T. B. K.; Wong, G. S. K.
The Journal of Organic Chemistry, 56, 872-872 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service