Skip to Content
Merck
All Photos(1)

Key Documents

520306

Sigma-Aldrich

2,4-Difluorobenzylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2ZnBr
CAS Number:
Molecular Weight:
272.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.995 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Fc1ccc(C[Zn]Br)c(F)c1

InChI

1S/C7H5F2.BrH.Zn/c1-5-2-3-6(8)4-7(5)9;;/h2-4H,1H2;1H;/q;;+1/p-1

InChI key

CMANCPNRVFBTQS-UHFFFAOYSA-M

Application

2,4-Difluorobenzylzinc bromide can be used to prepare nonpeptidomimetic dual XIAP and cIAP1 antagonists.[1]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a potent nonpeptidomimetic, small-molecule antagonist of cellular inhibitor of apoptosis protein 1 (cIAP1) and X-linked inhibitor of apoptosis protein (XIAP)
Tamanini E, et al.
Journal of medicinal chemistry, 60(11), 4611-4625 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service