Skip to Content
Merck
All Photos(1)

Key Documents

519715

Sigma-Aldrich

4-Propoxyaniline

Synonym(s):

4-Propoxyphenylamine, p-Propoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OC6H4NH2
CAS Number:
Molecular Weight:
151.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

SMILES string

CCCOc1ccc(N)cc1

InChI

1S/C9H13NO/c1-2-7-11-9-5-3-8(10)4-6-9/h3-6H,2,7,10H2,1H3

InChI key

DWOIGSLSPPLRKO-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of aggrecanase inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calix [4] arene with Lower-Rim ?-Amino a, ?-Unsaturated Ketones Containing Bis-Chelating Sites as a Highly Selective Fluorescence Turn-On Chemosensor for Two Copper (II) Ions.
Ho I, et al.
European Journal of Organic Chemistry, 8, 1472-1481 (2011)
Light and Temperature as Dual Stimuli Lead to Self-Assembly of Hyperbranched Azobenzene-Terminated Poly (N-isopropylacrylamide).
Huang W, et al.
Polymer, 8(5), 183-183 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service