Skip to Content
Merck
All Photos(1)

Key Documents

519200

Sigma-Aldrich

3,5-Dibenzyloxybenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2O)2C6H3CH2OH
CAS Number:
Molecular Weight:
320.38
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

78-81 °C (lit.)

SMILES string

OCc1cc(OCc2ccccc2)cc(OCc3ccccc3)c1

InChI

1S/C21H20O3/c22-14-19-11-20(23-15-17-7-3-1-4-8-17)13-21(12-19)24-16-18-9-5-2-6-10-18/h1-13,22H,14-16H2

InChI key

MHHXKZKHZMSINU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan E Wright et al.
Angewandte Chemie (International ed. in English), 53(13), 3409-3413 (2014-03-29)
Explorations into a series of different approaches for 9-membered carbocycle formation have afforded the first reported example of a 9-exo-dig ring closure via a Au(III)-promoted reaction between an alkyne and an aryl ring as well as several additional, unique Friedel-Crafts-type
B(C6F5)3: an efficient catalyst for reductive alkylation of alkoxy benzenes and for synthesis of triarylmethanes using aldehydes.
Chandrasekhar S, et al.
Tetrahedron Letters, 50(48), 6693-6697 (2009)
The structure and partial synthesis of imbricatine, a benzyltetrahydroisoquinoline alkaloid from the starfish Dermasterias imbricata.
Burgoyne DL, et al.
Canadian Journal of Chemistry, 69(1), 20-27 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service