Skip to Content
Merck
All Photos(2)

Key Documents

518093

Sigma-Aldrich

Ethyl (triphenylphosphoranylidene)pyruvate

95%

Synonym(s):

2-Ethoxycarbonyl-2-oxoethylidenetriphenylphosphorane, NSC 647346, [(Carboxycarbonyl)methylene]triphenylphosphorane ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€337.00

€337.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€337.00

About This Item

Linear Formula:
(C6H5)3P=CHCOCO2C2H5
CAS Number:
Molecular Weight:
376.38
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€337.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

reaction suitability

reaction type: C-C Bond Formation

mp

175 °C (dec.) (lit.)

functional group

ester
ketone
phosphine

SMILES string

CCOC(=O)C(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H21O3P/c1-2-26-23(25)22(24)18-27(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-18H,2H2,1H3

InChI key

OQLOPRPWPANPIE-UHFFFAOYSA-N

Application

Reactant for:
  • Wittig condensation
  • Chemoenzymic preparation of hexafluoroleucine and tetrafluoroleucine

Reactant for synthesis of:
  • Ahiral and chiral cyclic dehydro-α-amino acid derivatives through nucleophilic addition
  • Peptidyl α-keto-based inhibitors of cruzain
  • Deoxyfluoro-L-arabinofuranosyl triazole nucleoside derivatives with antiviral activity
  • Anti-human immunodificiency virus analogs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service