Skip to Content
Merck
All Photos(1)

Key Documents

517658

Sigma-Aldrich

Lutetium(III) acetylacetonate hydrate

≥99.9% trace metals basis

Synonym(s):

Lu(acac)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCHC(O)CH3]3Lu · xH2O
CAS Number:
Molecular Weight:
472.29 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.9% trace metals basis

form

solid

reaction suitability

core: lutetium
reagent type: catalyst

mp

158-163 °C (lit.)

SMILES string

[H]O[H].CC(=O)\C=C(\C)O[Lu](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Lu.H2O/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;;1H2/q;;;+3;/p-3/b3*4-3-;;

InChI key

HRUDKMKHQGBECZ-KJVLTGTBSA-K

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service