Skip to Content
Merck
All Photos(1)

Key Documents

514675

Sigma-Aldrich

Ethyl 3,5-difluorobenzoylformate

96%

Synonym(s):

Ethyl 3,5-difluorophenylglyoxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€137.00

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€137.00

About This Item

Linear Formula:
F2C6H3COCO2C2H5
CAS Number:
Molecular Weight:
214.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.482 (lit.)

bp

247.4 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

functional group

ester
fluoro
ketone

SMILES string

CCOC(=O)C(=O)c1cc(F)cc(F)c1

InChI

1S/C10H8F2O3/c1-2-15-10(14)9(13)6-3-7(11)5-8(12)4-6/h3-5H,2H2,1H3

InChI key

JPTHBXHXKOKMTE-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.0 °F - closed cup

Flash Point(C)

65.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service