Skip to Content
Merck
All Photos(3)

Key Documents

512753

Sigma-Aldrich

2-Chloro-6-methoxyphenylboronic acid

≥95%

Synonym(s):

2-Chloro-6-methoxybenzeneboronic acid, 6-Chloro-2-methoxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)B(OH)2
CAS Number:
Molecular Weight:
186.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

162-165 °C (lit.)

functional group

chloro

SMILES string

COc1cccc(Cl)c1B(O)O

InChI

1S/C7H8BClO3/c1-12-6-4-2-3-5(9)7(6)8(10)11/h2-4,10-11H,1H3

InChI key

XNWCIDBPLDKKAG-UHFFFAOYSA-N

Application

2-Chloro-6-methoxyphenylboronic acid can be used as a reactant:
  • In the palladium-catalyzed Suzuki-Miyaura coupling reaction.[1][2]
  • To synthesize canthin-6-one alkaloids by reacting with 8-bromo-1,5-naphthyridin-2-one via Pd-catalyzed Suzuki coupling and Cu-catalyzed amidation reactions.[2]
  • To prepare tryptamines via Suzuki coupling of vinylsulfonylmethyl resin-bound bromotryptamine.[3]

Reactant for:
  • Suzuki coupling
  • Preparation of inhibitors of cholesteryl ester transfer protein to raise HDLc levels
  • Pd-catalyzed Suzuki-Miyaura coupling
  • Solid-phase synthesis of 2,3,5-trisubstituted indoles

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-Step Total Syntheses of Canthin-6-one Alkaloids: New One-Pot Sequential Pd-Catalyzed Suzuki- Miyaura Coupling and Cu-Catalyzed Amidation Reaction
Gollner A and Koutentis PA
Organic Letters, 12(6), 1352-1355 (2010)
Versatile synthesis of functionalised dibenzothiophenes via Suzuki coupling and microwave-assisted ring closure
Rodriguez-Aristegui S, et al.
Organic & Biomolecular Chemistry, 9(17), 6066-6074 (2011)
A versatile linkage strategy for solid-phase synthesis of N, N-dimethyltryptamines and β-carbolines
Wu TYH and Schultz PG
Organic Letters, 4(23), 4033-4036 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service