Skip to Content
Merck
All Photos(1)

Documents

510874

Sigma-Aldrich

Tetraethylsilane

99%

Synonym(s):

Bis(pentamethylcyclopentadienyl)dimethylzirconium, Dimethylbis(pentamethylcyclopentadienyl)zirconium, Dimethylpermethylzirconocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(C2H5)4
CAS Number:
Molecular Weight:
144.33
Beilstein:
1732768
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.4263 (lit.)

bp

153-154 °C (lit.)

mp

−82.5 °C (lit.)

density

0.761 g/mL at 25 °C (lit.)

SMILES string

CC[Si](CC)(CC)CC

InChI

1S/C8H20Si/c1-5-9(6-2,7-3)8-4/h5-8H2,1-4H3

InChI key

VCZQFJFZMMALHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nashwa M Saleh et al.
Bioorganic chemistry, 104, 104350-104350 (2020-11-05)
Novel series of diazepam bearing sulfonamide moieties 5a-f and 7a-c were designed, synthesized and evaluated for anticancer activity against HepG2, HCT-116 and MCF-7 cell lines. MCF-7 was the most sensitive cell line to the influence of the new derivatives. In

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Synthesis of Melting Gels Using Mono-Substituted and Di-Substituted Alkoxysiloxanes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service