Skip to Content
Merck
All Photos(1)

Key Documents

50929

Sigma-Aldrich

Tridecafluorohexane-1-sulfonic acid potassium salt

≥98.0% (T)

Synonym(s):

1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonic acid potassium salt, PFHxS potassium salt, Perfluorohexanesulfonic acid potassium salt, Potassium tridecafluoro-1-hexanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6F13KO3S
CAS Number:
Molecular Weight:
438.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

solid

functional group

fluoro
sulfonic acid

SMILES string

[K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6HF13O3S.K/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22;/h(H,20,21,22);/q;+1/p-1

InChI key

RSCGQEBKFSGWJT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tridecafluorohexane-1-sulfonic acid potassium salt (PFHxS), also known as potassium perfluorohexanesulfonate, is a perfluorochemical (PFC) that belongs to the perfluoroalkyl sulfonates group. Its presence as a contaminant in the effluents of Taiwanese industrial wastewater treatment plant (IWWTP) and municipal wastewater treatment plants (MWWTPs) has been evaluated. It has been detected in the human serum samples by on-line solid-phase extraction (SPE) method coupled to high-performance liquid chromatography (HPLC)-tandem mass spectrometry (MS/MS).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanne Friis Berntsen et al.
Toxicology and applied pharmacology, 357, 19-32 (2018-08-26)
Perfluoroalkyl acids (PFAAs) are persistent compounds used in many industrial as well as consumer products. Despite restrictions, these compounds are found at measurable concentrations in samples of human and animal origin. In the present study we examined whether the effects
Zsuzsanna Kuklenyik et al.
Analytical chemistry, 77(18), 6085-6091 (2005-09-15)
We have developed an on-line solid-phase extraction (SPE) method coupled to high-performance liquid chromatography (HPLC)-tandem mass spectrometry (MS/MS) for measuring trace levels of 18 perfluorinated chemicals (3 perfluorosulfonates, 8 perfluorocarboxylates, 7 perfluorosulfonamides) in serum. Without protein precipitation, only dilution with
Angela Yu-Chen Lin et al.
Chemosphere, 80(10), 1167-1174 (2010-07-21)
We studied perfluorochemicals (PFCs) in the influents and effluents of two municipal wastewater treatment plants (MWWTPs) and in the effluents of an industrial wastewater treatment plant (IWWTP). The impact of IWWTP effluents on the receiving rivers (the Nanmen and Keya
Marisa Pfohl et al.
Toxicological sciences : an official journal of the Society of Toxicology, 178(2), 311-324 (2020-09-30)
Perfluoroalkyl substances (PFAS) represent a family of environmental toxicants that have infiltrated the living world. This study explores diet-PFAS interactions and the impact of perfluorooctanesulfonic acid (PFOS) and perfluorohexanesulfonic (PFHxS) on the hepatic proteome and blood lipidomic profiles. Male C57BL/6J
Kate M Annunziato et al.
Environmental health perspectives, 128(9), 97006-97006 (2020-09-24)
Drinking water contamination related to the use of aqueous film-forming foam (AFFF) has been documented at hundreds of military bases, airports, and firefighter training facilities. AFFF has historically contained high levels of long-chain per- and polyfluoroalkyl substances (PFAS), which pose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service