Skip to Content
Merck
All Photos(1)

Key Documents

499803

Sigma-Aldrich

2-Chlorophenylzinc iodide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4ZnI
CAS Number:
Molecular Weight:
303.84
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.973 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Clc1ccccc1[Zn]I

InChI

1S/C6H4Cl.HI.Zn/c7-6-4-2-1-3-5-6;;/h1-4H;1H;/q;;+1/p-1

InChI key

MADDWNUAYOHWRC-UHFFFAOYSA-M

Application

2-Chlorophenylzinc iodide is an organozinc compound, which can be used as a reactant in Negishi cross-coupling reaction to construct carbon-carbon bonds by reacting with organic halides using nickel or palladium catalysts.[1]
It can also be utilized as a reactant to prepare:
  • Arylmethanes via rhodium-catalyzed cross-coupling reaction with methyl halides.[2]
  • (2-chlorobenzyl)trimethylsilane by Rh-catalyzed cross-coupling reaction with (iodomethyl)trimethylsilane.[3]
  • Dichloro biphenyl by palladium catalyzed oxidative homocoupling reaction using N-chlorosuccinimide (NCS) or O2 as the oxidant.[4]

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of functionalized benzylsilanes from arylzinc compounds and (iodomethyl) trimethylsilane by means of a novel Rh catalysis
Takahashi H, et al.
The Journal of Organic Chemistry, 71(2), 671-675 (2006)
Palladium-catalyzed synthesis of biaryls from arylzinc compounds using N-chlorosuccinimide or oxygen as an oxidant
Hossain KM, et al.
Bulletin of the Chemical Society of Japan, 74(12), 2415-2420 (2001)
Novel Rh (I)-Catalyzed reaction of arylzinc compounds with methyl halides
T Kentaro, et al.
Chemistry Letters (Jpn), 1999(11), 1241-1242 (1999)
Mechanochemical Activation of Zinc and Application to Negishi Cross-Coupling
Cao Q, et al.
Angewandte Chemie (International Edition in English), 130(35), 11509-11513 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service