Skip to Content
Merck
All Photos(1)

Key Documents

498475

Sigma-Aldrich

4-(Ethoxycarbonyl)phenylzinc iodide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2CC6H4ZnI
CAS Number:
Molecular Weight:
341.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

1.018 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc([Zn]I)cc1

InChI

1S/C9H9O2.HI.Zn/c1-2-11-9(10)8-6-4-3-5-7-8;;/h4-7H,2H2,1H3;1H;/q;;+1/p-1

InChI key

QKMCGFIQAJUQIL-UHFFFAOYSA-M

Application

4-(Ethoxycarbonyl)phenylzinc iodide can be used as:
  • A substrate in the transition metal-catalyzed cross-coupling reactions with unsaturated thioethers and thiomethyl-substituted N-heterocycles.[1][2]
  • A starting material in the synthesis of indazole derived glucagon receptor antagonists.[3]
  • A substrate in the Pd-catalyzed synthesis of 5-aryl-2-furaldehydes by reacting with 5-bromo-2-furaldehyde.[4]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-catalyzed cross-coupling of functionalized organozinc reagents with thiomethyl-substituted heterocycles
Metzger A, et al.
Organic Letters, 11(18), 4228-4231 (2009)
Discovery of potent and orally bioavailable indazole-based glucagon receptor antagonists for the treatment of type 2 diabetes
Xu G, et al.
Bioorganic & Medicinal Chemistry Letters, 29(20), 126668-126668 (2019)
Pd-and Ni-Catalyzed Cross-Coupling Reactions of Functionalized Organozinc Reagents with Unsaturated Thioethers
Melzig L, et al.
Chemistry?A European Journal , 17(10), 2948-2956 (2011)
A convenient synthesis of 5-aryl-and 5-heteroaryl-2-furaldehydes by the cross-coupling reaction of organozincs
Kim S and Rieke RD
Tetrahedron Letters, 51(19), 2657-2659 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service