All Photos(1)
About This Item
Linear Formula:
CH3SO3C6H5
CAS Number:
Molecular Weight:
172.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
bp
279 °C (lit.)
mp
58-61 °C (lit.)
SMILES string
CS(=O)(=O)Oc1ccccc1
InChI
1S/C7H8O3S/c1-11(8,9)10-7-5-3-2-4-6-7/h2-6H,1H3
InChI key
WXVUCMFEGJUVTN-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Solutes in sulfuric acid. Part VIII. Protonation of phenol, 4-fluorophenol and the 2-and 4-sulfonates of anisole and phenol in concentrated aqueous sulfuric acid; determination of pKa values by means of 13C NMR.
Koeberg-Telder A, et al.
Rec. Trav. Chim., 102(6), 293-298 (1983)
Model studies on the photochemistry of phenolic sulfonate photoacid generators.
Andraos J, et al.
Chemistry of Materials, 10(6), 1694-1699 (1998)
Hiroshi Qadota et al.
Molecular biology of the cell, 29(17), 2084-2097 (2018-06-28)
Protein phosphatase 2A (PP2A) is a heterotrimer composed of single catalytic and scaffolding subunits and one of several possible regulatory subunits. We identified PPTR-2, a regulatory subunit of PP2A, as a binding partner for the giant muscle protein UNC-89 (obscurin)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service