Skip to Content
Merck
All Photos(3)

Key Documents

494968

Sigma-Aldrich

Thianaphthene-3-carboxaldehyde

95%

Synonym(s):

Benzo[b]thiophene-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6OS
CAS Number:
Molecular Weight:
162.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 494968, please contact your local Merck office or dealer. Contact Technical Service

Assay

95%

bp

166 °C/20 mmHg (lit.)

mp

53-57 °C (lit.)

functional group

aldehyde

SMILES string

O=Cc1csc2ccccc12

InChI

1S/C9H6OS/c10-5-7-6-11-9-4-2-1-3-8(7)9/h1-6H

InChI key

WDJLPQCBTBZTRH-UHFFFAOYSA-N

General description

Thianaphthene-3-carboxaldehyde, also known as benzo[b]thiophene-3-carboxaldehyde, can be synthesized from 3-methyl-benzo[b]thiophene.[1] It undergoes phosphine-free palladium coupling with aryl halides to form 2-arylbenzo[b]thiophenes.[2]

Application

Thianaphthene-3-carboxaldehyde (Benzo[b]thiophene-3-carboxaldehyde) may be used as a starting material in the multi-step synthesis of anthra[2,3-b:7,6-b′]bis[1benzothiophenes (ABBTs).[3]
It may be used in the synthesis of :
  • 6-(N,N-dimethylamino)-2-(benzo[b]thiophen-3-yl)quinazolin-4-one[4]
  • 6-(pyrrolidin-1-yl)-2-(benzo[b]thiophen-3-yl)quinazolin-4-one[4]
  • (Z)-2-(benzo­[b]­thio­phen-3-yl­methyl­ene)-1-aza­bi­cyclo­[2.2.2]­octan-3-one[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Hour et al.
British journal of pharmacology, 169(7), 1574-1586 (2013-05-04)
Our previous study demonstrated that 6-(pyrrolidin-1-yl)-2-(3-methoxyphenyl)quinazolin-4-one (HMJ38) was a potent anti-tubulin agent. Here, HMJ38 was used as a lead compound to develop more potent anti-cancer agents and to examine the anti-cancer mechanisms. Using computer-aided drug design, 2-aryl-6-substituted quinazolinones (MJ compounds)
An efficient phosphine-free palladium coupling for the synthesis of new 2-arylbenzo[b]thiophenes.
Chabert JFD, et al.
Tetrahedron, 60(14), 3221-3230 (2004)
Synthesis and properties of isomerically pure anthrabisbenzothiophenes.
Lehnherr D, et al.
Organic Letters, 14(1), 62-65 (2011)
Benzo[b]thiophene derivatives. VIII. Benzo[b]thiophene-3-earboxaldehyde and derivatives.
Campaigne E and Neiss ES.
Journal of Heterocyclic Chemistry, 3(1), 46-50 (1966)
(Z)-2-(Benzo[b] thiophen-3-ylmethylene)-1-azabicyclo [2.2.2] octan-3-one.
Sonar VN, et al.
Acta Crystallographica Section E, Structure Reports Online, 59(11), o1726-o1728 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service