Skip to Content
Merck
All Photos(1)

Key Documents

494119

Sigma-Aldrich

Deoxo-Fluor

Synonym(s):

Bis(2-methoxyethyl)aminosulfur trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3OCH2CH2)2NSF3
CAS Number:
Molecular Weight:
221.24
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

density

1.2 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COCCN(CCOC)S(F)(F)F

InChI

1S/C6H14F3NO2S/c1-11-5-3-10(4-6-12-2)13(7,8)9/h3-6H2,1-2H3

InChI key

APOYTRAZFJURPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For a review of recent work on nucleophilic fluorination of organic compounds with Deoxo-Fluor and DAST, see Synthesis, 2002, 17, 2561.[1]
One flask conversion of carboxylic acids to amides via the intermediacy of acyl fluorides.[2]

Legal Information

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 2561-2561 (2002)
Jonathan M White et al.
The Journal of organic chemistry, 69(7), 2573-2576 (2004-03-31)
The use of the Deoxo-Fluor reagent is a versatile method for acyl fluoride generation and subsequent one-flask amide coupling. It provides mild conditions and facile purification of the desired products in good to excellent yields. We have explored the utility

Articles

This technical article discusses Bis(2-methoxyethyl)aminosulfur Trifluoride (Deoxo-Fluor®).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service