Skip to Content
Merck
All Photos(1)

Key Documents

492825

Sigma-Aldrich

Iodomethane-d

≥98 atom % D, ≥99% (CP), contains copper as stabilizer

Synonym(s):

Methyl-d iodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€724.00

€724.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 G
€724.00

About This Item

Linear Formula:
DCH2I
CAS Number:
Molecular Weight:
142.95
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

€724.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.526 (lit.)

bp

42 °C (lit.)

mp

-66.5 °C (lit.)

density

2.296 g/mL at 25 °C

application(s)

agriculture
environmental
pharmaceutical

mass shift

M+1

storage temp.

2-8°C

SMILES string

[2H]CI

InChI

1S/CH3I/c1-2/h1H3/i1D

InChI key

INQOMBQAUSQDDS-MICDWDOJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.8 °F - closed cup

Flash Point(C)

32.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeneng Wang et al.
Nature, 472(7341), 57-63 (2011-04-09)
Metabolomics studies hold promise for the discovery of pathways linked to disease processes. Cardiovascular disease (CVD) represents the leading cause of death and morbidity worldwide. Here we used a metabolomics approach to generate unbiased small-molecule metabolic profiles in plasma that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service