Skip to Content
Merck
All Photos(1)

Key Documents

491233

Sigma-Aldrich

N-Methyl-d3-pyrrolidine

98 atom % D

Synonym(s):

1-Methyl-d3-pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€773.00

€773.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€773.00

About This Item

Empirical Formula (Hill Notation):
C5D3H8N
CAS Number:
Molecular Weight:
88.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€773.00


Please contact Customer Service for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

refractive index

n20/D 1.427 (lit.)

bp

80-81 °C (lit.)

density

0.848 g/mL at 25 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N1CCCC1

InChI

1S/C5H11N/c1-6-4-2-3-5-6/h2-5H2,1H3/i1D3

InChI key

AVFZOVWCLRSYKC-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal Ryszka et al.
The Journal of chemical physics, 147(9), 094303-094303 (2017-09-10)
Dissociative electron attachment to nicotine, pyridine, and N-methyl-pyrrolidine was studied in the gas phase in order to assess their stability with respect to low-energy electron interactions. Anion yield curves for different products at electron energies ranging from zero to 15

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service