Skip to Content
Merck
All Photos(1)

Key Documents

489646

Sigma-Aldrich

Potassium palmitate-1-13C

99 atom % 13C

Synonym(s):

Hexadecanoic acid-1-13C potassium salt, Palmitic acid-1-13C potassium salt, Potassium hexadecanoate-1-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€176.00

€176.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
€176.00

About This Item

Linear Formula:
CH3(CH2)1413CO2K
CAS Number:
Molecular Weight:
295.51
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€176.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C

form

solid

mp

285 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[K+].CCCCCCCCCCCCCCC[13C]([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i16+1;

InChI key

MQOCIYICOGDBSG-OMVBPHMTSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service