Skip to Content
Merck
All Photos(1)

Documents

489247

Sigma-Aldrich

Ethane-d5

99 atom % D

Synonym(s):

1,1,1,2,2-Pentadeuterioethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2H
CAS Number:
Molecular Weight:
35.10
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

vapor density

1.05 (vs air)

vapor pressure

37.95 atm ( 21.1 °C)

isotopic purity

99 atom % D

autoignition temp.

881 °F

bp

−88 °C (lit.)

mp

−172 °C (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])C([2H])([2H])[2H]

InChI

1S/C2H6/c1-2/h1-2H3/i1D2,2D3

InChI key

OTMSDBZUPAUEDD-KPAILUHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z512966 and Z512974 is recommended.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service