Skip to Content
Merck
All Photos(1)

Documents

487376

Sigma-Aldrich

3-Nitrophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NCS
CAS Number:
Molecular Weight:
180.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

57-60 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cccc(c1)N=C=S

InChI

1S/C7H4N2O2S/c10-9(11)7-3-1-2-6(4-7)8-5-12/h1-4H

InChI key

OEZXLKSZOAWNJU-UHFFFAOYSA-N

Related Categories

General description

3-Nitrophenyl isothiocyanate, also known as 1-isothiocyanato-3-nitrobenzene, is an organic building block containing an isocyanate group. Its enthalpy of vaporization at boiling point has been reported. Some of its physical properties like freezing point, boiling point, density and refractive index have been evaluated. Its synthesis from 3-nitroaniline has been reported.

Application

3-Nitrophenyl isothiocyanate may be used in the synthesis of 2-[(3-nitrophenyl)amino]naphtho[2,1-b]furo-5H-[3,2-d][1,3,4]thiadiazolo[3,2-a]pyrimidin- 5-one and 5-methyl-3-(3-nitrophenyl)-2-thiooxazolidin-4-one.
It may be used in the synthesis of the following thiourea derivatives:
  • N-[(3-chlorophenyl)methyl]-N′-(3-nitrophenyl)thiourea
  • N-[(5-chloro-2-methoxyphenyl)methyl]-N′-(3-nitrophenyl)thiourea
  • R/S-N-[6-chlorochroman-4-yl]-N′-(3-nitrophenyl)thiourea

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of novel naphtho[2,1-b]furo-5H-[3,2-d][1,3,4] thiadiazolo[3,2-a]pyrimidin-5-ones.
Ravindra KC, et al.
ARKIVOC (Gainesville, FL, United States), 11(1) (2008)
N-Aryl-N'-(chroman-4-yl) ureas and thioureas display in vitro anticancer activity and selectivity on apoptosis-resistant glioblastoma cells: Screening, synthesis of simplified derivatives, and structure-activity relationship analysis.
Goffin E, et al.
European Journal of Medicinal Chemistry, 54, 834-844 (2012)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 536-536 (2014)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 138-138 (2015)
A highly efficient methodology for 5-methyl-3-aryl-2-thiooxazolidin-4-ones using lithium perchlorate in DIPEA mediated synthesis.
Khatik GL, et al.
Journal of Heterocyclic Chemistry, 47(3), 734-734 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service