Skip to Content
Merck
All Photos(1)

Key Documents

481947

Sigma-Aldrich

Phosphonitrilic chloride trimer

99.95% trace metals basis

Synonym(s):

1,3,5,2,4,6-Triazatriphosphorine-2,2,4,4,6,6-hexachloride, 1,3,5-Triaza-2,4,6-triphosphorin-2,2,4,4,6,6-hexachloride, Hexachlorocyclotriphosphazene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NPCl2)3
CAS Number:
Molecular Weight:
347.66
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace metals basis

mp

112-115 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

SMILES string

Clp1(Cl)np(Cl)(Cl)np(Cl)(Cl)n1

InChI

1S/Cl6N3P3/c1-10(2)7-11(3,4)9-12(5,6)8-10

InChI key

UBIJTWDKTYCPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the synthesis of ″dandelion″ (spherical) dendrimers. Ring-opening polymerization, ligand and/or ligand precursor for transition metals, and the study of P-Cl bond substitution reactions are among the interesting uses for this product.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Yeon Seong et al.
International journal of pharmaceutics, 314(1), 90-96 (2006-03-22)
Biocompatible and thermosensitive poly(organophosphazenes) with a lower critical solution temperature (LCST) below body temperature have been designed with the aim for the local delivery of peptide and protein drugs. These polymers could be synthesized by introducing short chain tri- or
Studies on the reaction of hexachlorocyclotriphosphazene and 2-hydroxyethyl methacrylate and on the physical properties of its polymer.
M Anzai et al.
The Journal of Nihon University School of Dentistry, 26(2), 109-118 (1984-06-01)
Tam T T Bui et al.
Chirality, 17(8), 438-443 (2005-08-13)
The gem-disubstituted cyclotriphosphazene 1 reacted with piperazine (pip) to give the piperazine-bridged derivative 2, which is expected to exist in meso and racemic forms because the two PCl (pip) groups are stereogenic. The proton-decoupled (31)P NMR spectrum of 2 gave
S Paasch et al.
Solid state nuclear magnetic resonance, 4(5), 267-280 (1995-07-01)
The present study deals with solid-state nuclear magnetic resonance (NMR) investigations on hexachlorocyclotriphosphazene (N3P3Cl6) and T and K form of octachlorocyclotetraphosphazene (N4P4Cl8). Using 31P NMR we have recorded spectra of different magic-angle spinning (MAS) frequencies and static powder spectra. By
David K Y Lee et al.
Dalton transactions (Cambridge, England : 2003), 39(22), 5341-5348 (2010-05-05)
Two novel cyclic phosphazenes with asymmetric spiro rings were synthesized via reactions of hexachlorocyclotriphosphazene with chiral amino alcohol residues. The reactions showed preferential formation of the cis isomer possibly due to the delocalization of the lone pair electrons of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service