Skip to Content
Merck
All Photos(1)

Key Documents

480584

Sigma-Aldrich

1-Cyclohexenyl trifluoromethanesulfonate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€202.00

€202.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€202.00

About This Item

Empirical Formula (Hill Notation):
C7H9F3O3S
CAS Number:
Molecular Weight:
230.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€202.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

bp

84-87 °C/4 mmHg (lit.)

density

1.315 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

storage temp.

2-8°C

SMILES string

FC(F)(F)S(=O)(=O)OC1=CCCCC1

InChI

1S/C7H9F3O3S/c8-7(9,10)14(11,12)13-6-4-2-1-3-5-6/h4H,1-3,5H2

InChI key

WVSCRRLWRRANJY-UHFFFAOYSA-N

General description

1-Cyclohexenyl trifluoromethanesulfonate, also known as 1-cyclohexenyl triflate, is a cyclohexenyl sulfonate. Its trifluoromethylation reaction in the presence of different monodentate biaryl phosphine ligands has been investigated.[1] The asymmetric Heck reaction of 1-cyclohexenyl trifluoromethanesulfonate using palladium complexes of phosphine oxazoline ligand has been studied.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

160.0 °F - closed cup

Flash Point(C)

71.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proline derived phosphine-oxazoline ligands in the asymmetric Heck reaction.
Gilbertson SR, et al.
Tetrahedron Letters, 42(3), 365-368 (2001)
Eun Jin Cho et al.
Organic letters, 13(24), 6552-6555 (2011-11-25)
A method for the palladium-catalyzed trifluoromethylation of cyclohexenyl sulfonates has been developed. Various cyclohexenyl triflates and nonaflates underwent trifluoromethylation under mild reaction conditions using a catalyst system composed of Pd(dba)(2) or [(allyl)PdCl](2) and the monodentate biaryl phosphine ligand (t)BuXPhos. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service