Skip to Content
Merck
All Photos(1)

Key Documents

480037

Sigma-Aldrich

Magtrieve

Synonym(s):

Chromium dioxide, Chromium dioxide (CrO2 ), Chromium oxide, Chromium(IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrO2
CAS Number:
Molecular Weight:
83.99
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

>375 °C (dec.) (lit.)

density

4.85 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Cr]=O

InChI

1S/Cr.2O

InChI key

AYTAKQFHWFYBMA-UHFFFAOYSA-N

Application

Facilitates high yield aromatization of Hantzsch 1,4-dihydropyridines to the corresponding pyridines.[1]

Legal Information

DuPont Product ®Reg. trademark of E.I. du Pont de Nemours & Co., Inc.
Magtrieve is a trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Feng et al.
Metabolism: clinical and experimental, 50(10), 1168-1174 (2001-10-05)
In order to study the metabolism of essential trace elements in diabetics, we studied alloxan-diabetic rats for the distribution patterns of chromium (Cr), cobalt (Co), iron (Fe), selenium (Se), and zinc (Zn) in the liver, kidney, pancreas, and testes, as
Ko, K-Y. Kim, J-Y.
Tetrahedron Letters, 40, 3207-3207 (1999)
Immobilization of antigens and antibodies on chromium dioxide magnetic particles for use in immunodiagnostic assays.
H P Lau et al.
Targeted diagnosis and therapy, 2, 201-216 (1989-01-01)
K P Lee et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 10(1), 125-145 (1988-01-01)
Rats were exposed by inhalation to chromium dioxide (CrO2) dust at design concentrations of 0, 0.5 mg/m3 (stabilized and unstabilized, respectively), or 25 mg/m3 (stabilized) for 6 hr/day, 5 days/week for 2 years. No dust-exposure-related pathological changes other than lung
Plasmodium berghei: in vivo efficacy of albendazole in different rodent models.
G S Dow et al.
Experimental parasitology, 88(2), 154-156 (1998-04-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service