Skip to Content
Merck
All Photos(1)

Key Documents

476455

Sigma-Aldrich

Isopropyl formate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€83.60

€83.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€83.60

About This Item

Linear Formula:
HCO2CH(CH3)2
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€83.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.368 (lit.)

bp

68 °C (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)OC(C)C

InChI

1S/C4H8O2/c1-4(2)6-3-5/h3-4H,1-2H3

InChI key

RMOUBSOVHSONPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

7.0 °F - closed cup

Flash Point(C)

-13.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reactions of alkyl radicals. Part 2.-Methyl radical photosensitized decomposition of n-propyl and isopropyl formates.
Thynne JCJ.
Transactions of the Faraday Society, 58, 1394-1402 (1962)
The kinetics of the pyrolysis of ethyl and isopropyl formates and acetates.
Blades AT.
Canadian Journal of Chemistry, 32(4), 366-372 (1954)
Conformational analysis and vibrational spectra of isopropyl formate and some deuterated isotopomers.
Rosas RL, et al.
Journal of Raman Spectroscopy, 24(3), 143-165 (1993)
Atmospheric chemistry of isopropyl formate and tert-butyl formate.
Pimentel AS, et al.
International Journal of Chemical Kinetics, 42(8), 479-498 (2010)
Low resolution microwave spectroscopy, part IV: The conformation of isopropyl formate, isopropyl fluoroformate, isopropyl chloroformate, isopropyl cyanoformate, and isopropyl trifluoroacetate.
True NS and Bohn RK.
Journal of Molecular Structure, 36(2), 173-181 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service