Skip to Content
Merck
All Photos(1)

Key Documents

475955

Sigma-Aldrich

3-Methoxyphenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NCS
CAS Number:
Molecular Weight:
165.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.64 (lit.)

bp

129 °C/11 mmHg (lit.)

density

1.179 g/mL at 25 °C (lit.)

functional group

isothiocyanate

SMILES string

COc1cccc(c1)N=C=S

InChI

1S/C8H7NOS/c1-10-8-4-2-3-7(5-8)9-6-11/h2-5H,1H3

InChI key

WHBYCPUKGYEYFU-UHFFFAOYSA-N

General description

3-Methoxyphenyl isothiocyanate, also referred as m-methoxyphenyl isothiocyanate,[1] is an iosthiocyanate derivative.

Application

3-Methoxyphenyl isothiocyanate may be used in the synthesis of the following:
  • 2-[(3-methoxyphenyl)amino]naphtho[2,1-b]furo-5H-[3,2-d][1,3,4]thiadiazolo[3,2-a]pyrimidin-5-one[2]
  • N-2 pyridyl-N′-3-methoxyphenylthioureas[3]
  • acylthiosemicarbazides[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural, thermal and spectral studies of N-2-pyridyl-, N-2-picolyl-and N-2-(4, 6-lutidyl)-N'-(3-methoxyphenyl) thioureas.
Giesen JM, et al.
Journal of Molecular Structure, 613(1), 223-233 (2002)
Yalkowsky SH, et al.
Handbook of Aqueous Solubility Data, 465-465 (2010)
Acton QA.
Issues in Chemistry and General Chemical Research, 170-170 (2013)
Synthesis and antimicrobial activity of novel naphtho [2, 1-b] furo-5H-[3, 2-d][1, 3, 4] thiadiazolo [3, 2-a] pyrimidin-5-ones.
Ravindra KC, et al.
ARKIVOC (Gainesville, FL, United States), 11(1) (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service