Skip to Content
Merck
All Photos(1)

Key Documents

475335

Sigma-Aldrich

1,4-Dimethoxy-2-butyne

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2C≡CCH2OCH3
CAS Number:
Molecular Weight:
114.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.434 (lit.)

bp

70 °C/30 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COCC#CCOC

InChI

1S/C6H10O2/c1-7-5-3-4-6-8-2/h5-6H2,1-2H3

InChI key

YFUQYYGBJJCAPR-UHFFFAOYSA-N

General description

1,4-Dimethoxy-2-butyne is a symmetrical aliphatic alkyne. Its reaction with Hg(OAc)2 (Mercury(II)acetate) has been investigated.

Application

1,4-Dimethoxy-2-butyne may be used in the preparation of alkyne complexes: decacarbonyl(μ3;-η2-verbar-1,4-dimethoxy-2-butyne)triiron and nonacarbonyl(μ3;-η2;-⊥-1,4-dimethoxy-3-butyne)triiron. It may also be used to synthesize benzonaphthosiline.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Decacarbonyl (?3;-η2-ll-1, 4-dimethoxy-2-butyne) triiron, an Iron Cluster with Unusual Structure.
Lentz D and Reuter M.
Chemische Berichte, 124(4), 773-775 (1991)
Metalation of alkynes. 1. Effect of alkyne structure on the rate of acetoxymercuration.
Bassetti M and Floris B.
The Journal of Organic Chemistry, 51(22), 4140-4143 (1986)
Ryo Shintani et al.
Chemical communications (Cambridge, England), 51(57), 11378-11381 (2015-06-19)
Rhodium-catalyzed intramolecular alkynylsilylation of alkynes is described. The reaction proceeds through syn-insertion by a cationic rhodium/triarylphosphine catalyst, representing the first alkynylsilylation of alkynes via the cleavage of a C(sp)-Si bond by transition-metal catalysis. A highly enantioselective variant is also described
One-pot synthesis of isoquinolinium salts by rhodium-catalyzed C-H bond activation: application to the total synthesis of oxychelerythrine.
Jayachandran Jayakumar et al.
Angewandte Chemie (International ed. in English), 51(1), 197-200 (2011-11-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service