Skip to Content
Merck
All Photos(1)

Documents

47475

Sigma-Aldrich

Fmoc-Pro-OPfp

≥96.0%

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-proline pentafluorophenyl ester, Fmoc-L-proline pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18F5NO4
CAS Number:
Molecular Weight:
503.42
Beilstein:
4731096
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥96.0%

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

127-129 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(OC(=O)[C@@H]2CCCN2C(=O)OCC3c4ccccc4-c5ccccc35)c(F)c1F

InChI

1S/C26H18F5NO4/c27-19-20(28)22(30)24(23(31)21(19)29)36-25(33)18-10-5-11-32(18)26(34)35-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2/t18-/m0/s1

InChI key

CQBLOHXKGUNWRV-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service