Skip to Content
Merck
All Photos(1)

Key Documents

473006

Sigma-Aldrich

(1S,4S)-(+)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide

97%

Synonym(s):

(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2 · 2HBr
CAS Number:
Molecular Weight:
259.97
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]23/D +22°, c = 1 in H2O

mp

300 °C (dec.) (lit.)

SMILES string

Br.Br.C1N[C@@H]2CN[C@H]1C2

InChI

1S/C5H10N2.2BrH/c1-4-2-6-5(1)3-7-4;;/h4-7H,1-3H2;2*1H/t4-,5-;;/m0../s1

InChI key

ISYQWKOXKGJREA-RSLHMRQOSA-N

Application

(1S,4S)-(+)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide can be used as:
  • A starting material for the synthesis of chiral diazabicyclic ligands, applicable in the preparation of dicopper(II) complexes.
  • A precursor in the preparation of diazabicyclo[2.2.1]heptane derivatives as catalysts, which are applicable in the asymmetric catalysis reactions.
  • An organocatalyst in Biginelli reaction of aromatic aldehydes with ethyl acetoacetate and urea.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of (1S, 4S)-2, 5-diazabicyclo [2.2. 1] heptane derivatives in asymmetric organocatalysis: the Biginelli reaction
Gonzalez-Olvera R, et al.
ARKIVOC (Gainesville, FL, United States), 6(4) (2008)
Dicopper (II) complexes of chiral C2-symmetric diamino-bis (2-methylpyridyl) and diamino-bis (2-methylbenzimidazolyl) ligands
Perez V, et al.
Inorganic Chemistry Communications, 14(2), 389-391 (2011)
Dicopper (II) complexes of chiral C2-symmetric diamino-bis (2-methylpyridyl) and diamino-bis (2-methylbenzimidazolyl) ligands
Melgar-Fernandez R, et al.
European Journal of Organic Chemistry, 2008(4), 655-672 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service