Skip to Content
Merck
All Photos(1)

Key Documents

472972

Sigma-Aldrich

Methyl (R)-(+)-2-(4-hydroxyphenoxy)propionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OC6H4OH)CO2CH3
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]22/D +43°, c = 1 in chloroform

mp

64-67 °C (lit.)

SMILES string

COC(=O)[C@@H](C)Oc1ccc(O)cc1

InChI

1S/C10H12O4/c1-7(10(12)13-2)14-9-5-3-8(11)4-6-9/h3-7,11H,1-2H3/t7-/m1/s1

InChI key

UUYSCNGPNOYZMC-SSDOTTSWSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service