Skip to Content
Merck
All Photos(1)

Documents

472190

Sigma-Aldrich

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CF3)NCO
CAS Number:
Molecular Weight:
205.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.455 (lit.)

bp

53 °C/2.5 mmHg (lit.)

density

1.418 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(cc1N=C=O)C(F)(F)F

InChI

1S/C8H3F4NO/c9-6-2-1-5(8(10,11)12)3-7(6)13-4-14/h1-3H

InChI key

NAIKHCBDZGSGHH-UHFFFAOYSA-N

General description

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate is a fluorinated building block.

Application

Useful pharmaceutical building block for the preparation of ureas from amines.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiqin Ji et al.
Bioorganic & medicinal chemistry letters, 16(16), 4326-4330 (2006-06-01)
A series of isothiazolopyrimidines and isoxazolopyrimidines were synthesized and identified as potent KDR inhibitors. SAR studies led to isothiazolopyrimidine urea analogs that potently inhibit VEGFR tyrosine kinases (KDR enzymatic and cellular IC(50) values below 10 nM) as well as cKIT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service