Skip to Content
Merck
All Photos(2)

Key Documents

467332

Sigma-Aldrich

Holmium(III) acetate hydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3Ho · xH2O
CAS Number:
Molecular Weight:
342.06 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

powder, crystals or chunks

composition

Degree of hydration, ~4

reaction suitability

core: holmium
reagent type: catalyst

SMILES string

O.CC(=O)O[Ho](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Ho.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

LCCGYXRQAXROOW-UHFFFAOYSA-K

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qiyue Shao et al.
Nanoscale, 9(33), 12132-12141 (2017-08-15)
Upconversion nanoparticles (UCNPs) are an excellent choice to construct security features against counterfeiting, owing to their unique NIR-to-VIS upconversion luminescence (UCL) characteristics. However, the application of upconversion materials is limited, due to their single and invariant emission colors. Herein, the
Rong Huang et al.
Nanoscale, 13(9), 4812-4820 (2021-02-27)
Photon upconversion is a fascinating phenomenon that can convert low-energy photons to high-energy photons efficiently. However, most previous relevant research has been focused on upconversion systems with a sufficiently low lanthanide emitter concentration, such as 2 mol% for Er3+ in
Bo Zhou et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700667-1700667 (2018-03-30)
Upconverting materials have achieved great progress in recent years, however, it remains challenging for the mechanistic research on new upconversion strategy of lanthanides. Here, a novel and efficient strategy to realize photon upconversion from more lanthanides and fine control of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service