Skip to Content
Merck
All Photos(1)

Key Documents

465585

Sigma-Aldrich

1,3-Dichloro-1,1,3,3-tetramethyldisiloxane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2Si(Cl)OSi(Cl)(CH3)2
CAS Number:
Molecular Weight:
203.21
Beilstein:
1741218
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.407 (lit.)

bp

138 °C (lit.)

density

1.039 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)O[Si](C)(C)Cl

InChI

1S/C4H12Cl2OSi2/c1-8(2,5)7-9(3,4)6/h1-4H3

InChI key

DMEXFOUCEOWRGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dichloro-1,1,3,3-tetramethyldisiloxane can be used to prepare:
  • A Silicon-based fluorene polymer by reacting with a dibromofluorene derived Grignard reagent.
  • ansa-Metallocene compound i.e, ansa-[TiCl255-(C5H4SiMe2)2O}].
  • Silaanthracenophanes.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon-Silicon coupling between fluorenyl Grignard reagents and dichlorosilanes for synthesis of novel copolymers and chemical properties of the copolymers
Kitamura N and Yamamoto T
Applied Organometallic Chemistry, 17(11), 840-842 (2003)
Synthesis and Structures of New Silaanthracenophanes
Lee I, et al.
Bulletin of the Korean Chemical Society,, 33(1), 255-260 (2012)
Synthesis and crystal structures of μ-1, 3-bis (η5-tetramethylcyclopentadienyl)-1, 1, 3, 3-tetramethyldisiloxane)[ η2-bis (trimethylsilyl) ethyne] titanium (II) and a doubly tucked-in product of its thermolysis
Horacek M, et al.
Journal of Organometallic Chemistry, 658(1-2), 235-241 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service