Skip to Content
Merck

Skip To

463582

Titanium(IV) methoxide

≥99.99% trace metals basis

Synonym(s):

Methyl orthotitanate, Tetramethoxytitanium, Tetramethyl orthotitanate, Tetramethyl titanate, Titanium tetramethoxide

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Ti(OCH3)4
CAS Number:
Molecular Weight:
172.00
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
EC Number:
213-596-4
MDL number:


Quality Segment

assay

≥99.99% trace metals basis

form

powder

reaction suitability

core: titanium, reagent type: catalyst

mp

200-210 °C (lit.)

SMILES string

CO[Ti](OC)(OC)OC

InChI

1S/4CH3O.Ti/c4*1-2;/h4*1H3;/q4*-1;+4

InChI key

ITNVWQNWHXEMNS-UHFFFAOYSA-N

General description

Atomic number of base material: 22 Titanium

Features and Benefits

Used for the preparation of polyoxotitanates.


Still not finding the right product?

Explore all of our products under Titanium(IV) methoxide


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

57.2 °F - closed cup

flash_point_c

14 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Reactive silicone chemistry: Focus on pure silicon production, polymerizations, and controlled stereochemistry reactions.

View All Articles




Global Trade Item Number

SKUGTIN
463582-25G04061833606780

Questions

Reviews

No rating value

Active Filters