Skip to Content
Merck
All Photos(1)

Key Documents

460656

Sigma-Aldrich

mono-Benzyl malonate

mono-Benzyl malonate
1 of 1 reviewers received a sample product or took part in a promotion

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€18.80

€18.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€18.80

About This Item

Linear Formula:
C6H5CH2O2CCH2CO2H
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€18.80


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

solid

mp

47-51 °C (lit.)

functional group

carboxylic acid
ester
phenyl

SMILES string

OC(=O)CC(=O)OCc1ccccc1

InChI

1S/C10H10O4/c11-9(12)6-10(13)14-7-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)

InChI key

CFLAHQSWDKNWPW-UHFFFAOYSA-N

General description

mono-Benzyl malonate can be synthesized by the reaction of Meldrum′s acid (2,2-dimethyl- 1,3-dioxane-4,6-dione) with benzyl alcohol.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A potential suicide inhibitor of adenylosuccinate lyase.
Hartgers A, et al.
Nucleosides and Nucleotides, 11(2), 1325-1332 (1992)

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Collegeville , PA
    • Review 1
    • Votes 0
    5 out of 5 stars.

    High purity

    The compound was pure and saved under inert gas which is nice.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service