Skip to Content
Merck
All Photos(1)

Key Documents

460486

Sigma-Aldrich

Isooctyl tallate

technical grade

Synonym(s):

Isooctyl esters tall-oil fatty acids

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12162002

grade

technical grade

refractive index

n20/D 1.461 (lit.)

bp

>316 °C (lit.)

solubility

hexane and toluene: soluble
water and propylene glycol: insoluble

density

0.87 g/mL at 25 °C (lit.)

Application

Plasticizer

Physical properties

High oleic ester content

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service